ATEX-Rated Solenoid Valves
ATEX-rated explosion-proof coils are designed for petrochemical and other industrial applications
  • Two way normally closed valves offer port sizes for many applications
  • Body material of 316 stainless steel withstands costic media
  • Max operating pressure of 230 psi withstands high-pressure applications
$543.00 - $1,061.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cv Orifice (in) Port Size (NPT(F))
Availability Pricing
ODE 21IN6KOV250-IH Solenoid Valve, 316 SS, with 3/8 NPT(F) Port EW-08610-40 Mfr # 21IN3KIV150-IH
COMPARE
10.57 0.63 0.375
$543.00USD / Each
ODE 21IN3KOV150-IH Solenoid Valve, 316 SS, with 1/2 NPT(F) Port EW-08610-42 Mfr # 21IN4KIV160-IH 2
COMPARE
13.21 0.63 0.5
$594.00USD / Each
ODE 21IN4KOV160-IH Solenoid Valve, 316 SS, with 3/4 NPT(F) Port EW-08610-44 Mfr # 21IN5KIV200-IH
COMPARE
15.85 0.79 0.75
$591.00USD / Each
ODE 21IN5KOV200-IH Solenoid Valve, 316 SS, with 1 NPT(F) Port EW-08610-46 Mfr # 21IN6KIV250-IH
COMPARE
36.98 0.98 1
$688.00USD / Each
ODE 21IN7KOV350-IH Solenoid Valve, 316 SS, with 1 1/4 NPT(F) Port EW-08610-48 Mfr # 21IN7KOV350-IH
COMPARE
79.25 1.38 1.25
$1,021.00USD / Each
ODE 21IN8KOV400-IH Solenoid Valve, 316 SS, with 1 1/2 NPT(F) Port EW-08610-50 Mfr # 21IN8KOV400-IH
COMPARE
89.82 1.57 1.5
$1,061.00USD / Each