This product is now discontinued, View Related Products
Automation USA Coating Thickness Gauge
Compact and robust, ideal for indoor and outdoor use. Readings appear on the digital display along with a substrate indicator. Designed for various applications such as assessing paintwork on an automobile, inspecting outdoor structures such as bridges or tanks or using on liquid or powder line. FEATURES
  • No calibration required
  • Automation ON/OFF switch
  • Precision and accuracy over complete measurement range
  • Hall effect technology for measurements in the hot, cold or wet climate
  • Measurements over steel (QualNix4200) or steel and aluminum (QualNix4500)
  • Compact, rugged and robust
  • Audible signal confirms measurement
  • Readings in both mils and um (microns) selectable
  • One hand operation for all applications
  • Durable integrated ruby sensor
  • Low battery indicator warning
  • Substrate indicator
  • Complies to national international (DIN, ISO, BS, ASTM) standards and regulations
    DIN 50981, 50984
    ISO 2178, 2360, 2808
    BS 5411 (3, 11), 3900 (C, 5)
    ASTM B499, D1400, D1186
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Range
Availability Pricing
TestCoat 315254-QNIX4200 Coating Thicknes Gauge, Fe 0/120mils 0/3000microns EW-59784-16 Mfr # 315254-QNIX4200
COMPARE
Fe: 0 to 120 mils/0 to 3000 microns
Call For Price
TestCoat 380000-QNIX4500 Coating Thickness Gauge; Ferrous/Non-Ferrous EW-59784-17 Mfr # 380000-QNIX4500
COMPARE
Fe: 0 to 120 mils/0 to 3000 microns; Non Fe: 0 to 80 mils/0 to 2000 microns