This product is now discontinued, View Related Products
Automation USA Ferrous - Non-Ferrous Coating Thickness Meter
This combination gauge, with its two integrated probes, non-destructively measures coatings on both ferrous and non-ferrous substrates.
There is no on/off switch. The special design of the instrument, with its duplex display and 10-second retention of the last-measured value, facilitates measurements in hard-to-reach corners.
The gauge is equipped with a non-wearing ruby probe-tip to ensure long life and precision of measurement. The “V”-groove facilitates measurements on rods and other curved surfaces. The unit is powered by a 9 Volt battery.
The Deluxe Model also offers memory, block segmentation, instant averaging and RS-232 download capability. Software and patchcord come included with this gauge which stores up to 3900 measurements, blocks the data and provides an average thickness right on the display. The Deluxe Model downloads the measurements to a menu-guided software program that lists measurements, provides statistics and displays a histogram.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Memory Output
Availability Pricing
TestCoat Coating Thickness Gauge, Ferrous/Nonferrous EW-59784-18 Mfr # 315056-QNIX1500
COMPARE
None None
Call For Price
TestCoat Coat Thickness Gauge w/RS-232 Memory Software EW-59784-19 Mfr # 315056-M-QNIX1500M
COMPARE
3900 readings RS-232