This product is now discontinued, View Related Products
B&K Precision 9115 Series Programmable DC Power Supplies
Replace several limited power supplies with a single, versatile high power supply
  • Multi-ranging operation automatically calculates voltage or current limits when one parameter is set for foolproof programming
  • Program and save up to 100 settings on internal memory to quickly recall common test sequences
  • Remote control by USB (TMC-compliant), RS-232, GPIB, RS-485, and analog signals allow power supply to easily integrate into existing systems
  • Master/slave operation allows connection of multiple supplies in series or parallel to increase voltage or current output
  • Adjustable voltage slope function simulates changing voltage conditions expected for certain Devices Under Test (DUTs) to experience
  • Overvoltage, overpower, and overtemperature protections, and key-lock function increase safety and security
  • Compact unit fits in a 1U rack to save space
Software is available for download from the manufacturer's website. It emulates the front panel, generates and executes test sequences, and logs measurement data to the PC. The software also integrates with LabVIEW apps for mobile devices (iOS, Android, Windows 8), allowing remote monitoring.
3 PRODUCT OPTIONS
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
B&K Precision 9115 Programmable DC Power Supply, 1200W EW-20019-90 Mfr # 9115
COMPARE
0 to 60 0 to 80 VDC ≤ 60 mVp-p; ≤ 100 mArms
Call For Price
B&K Precision 9115-AT Programmable DC Power Supply, 1200W, automotive tests EW-20019-91 Mfr # 9115-AT
COMPARE
0 to 60 0 to 80 VDC ≤ 60 mVp-p; ≤ 100 mArms
Call For Price
B&K Precision 9116 Programmable DC Power Supply, 1200W, 150 V/30 A EW-20019-92 Mfr # 9116
COMPARE
0 to 30 A 0 to 150 VDC ≤ 60 mVp-p; ≤ 50 mArms