This product is now discontinued, View Related Products
B&K Precision 9120 Series Single Output Programmable DC Power Supplies
Clean reliable power source is ideal for R & D design verification, production testing, or lab applications
  • Excellent display resolution
  • Fast transient response time (< 150 µs)
  • SCPI compatible
  • RS-232 interface (standard), USB optional
  • Closed case calibration
  • Compact size for bench use or rack mountable (2U x 1/2U size)
  • Digital voltmeter and milliohm meter
  • Discrete Fault Indicator/Remote Inhibit (DFI/RI) for turning multiple power supplies On/Off simultaneously
  • Included application software for front panel emulation and simple test sequence generation
  • Sheathed banana plug terminals for safety
5 PRODUCT OPTIONS
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
B&K Precision 9120A Single Output Programmable DC Power Supply, 96W EW-20036-70 Mfr # 9120A
COMPARE
0 to 3 A 0 to 32 VDC ≤ 4 mVp-p; ≤ 3 mArms
Call For Price
B&K Precision 9121A Single Output Programmable DC Power Supply, 100W EW-20036-71 Mfr # 9121A
COMPARE
0 to 5 A 0 to 20 VDC ≤ 3 mVp-p; ≤ 3 mArms
Call For Price
B&K Precision 9122A Single Output Programmable DC Power Supply, 150W EW-20036-72 Mfr # 9122A
COMPARE
0 to 2.5 A 0 to 60 VDC ≤ 5 mVp-p; ≤ 3 mArms
Call For Price
B&K Precision 9123A Single Output Programmable DC Power Supply, 150W EW-20036-73 Mfr # 9123A
COMPARE
0 to 5 A 0 to 30 VDC ≤ 4 mVp-p; ≤ 4 mArms
Call For Price
B&K Precision 9124 Single Output Programmable DC Power Supply, 86W EW-20036-74 Mfr # 9124
COMPARE
0 to 1.2 A 0 to 72 VDC ≤ 5 mVp-p; ≤ 3 mArms