This product is now discontinued, View Related Products
B&K Precision Programmable Switching DC Power Supplies
High power output at a low cost
  • RS-232 interface for remote control capability
  • Data logging capability
  • Output disable functionality
  • Overvoltage protection
  • Constant voltage and current operation (current limiting)
  • Large easy-to-read, 4-digit LCD
  • Optional RS-232 to RS-485 adapter for RS-485 applications
These programmable switching DC power supplies feature new switching technologies to offer more power at a lower cost than traditional linear power supplies. Units are ideal for repetitive test routines in R&D, production, product evaluation applications and more. The built-in micro-controller unit along with the included software lets you perform recalibration without opening the case. Use the built-in RS-232 interface to remotely control the power supply or program a sequence of different voltages and current limits that can be used in an automated test application. Units feature data logging with color graphic display in adjusting voltage range, current, watts, and time periods, providing valuable data analysis tools.
3 PRODUCT OPTIONS
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
B&K Precision 1696 Programmable Switching DC Power Supplies, 20 V, 10 A EW-20045-96 Mfr # 1696B
COMPARE
0 to 10 A 1 to 20 VDC ≤ 25 mVp-p; < 10 mArms
Call For Price
B&K Precision 1697 Programmable Switching DC Power Supplies, 40 V, 5 A EW-20045-97 Mfr # 1697
COMPARE
0 to 5 A 1 to 40 VDC ≤ 25 mVp-p; < 10 mArms
Call For Price
B&K Precision 1698 Programmable Switching DC Power Supplies, 60 V, 3.3 A EW-20045-98 Mfr # 1698
COMPARE
0 to 3.3 A 1 to 60 VDC ≤ 25 mVp-p; < 10 mArms