B&K Precision True RMS Bench Multimeters
Built-in math functions and USB connectivity enhance productivity
  • Dual display allowing multiple measurements to be conveniently displayed at once
  • Limit operation allows the setting and control of values that determine a HI / IN / LO status of subsequent measurements
  • AC volt & amp measurement over wide frequency range
  • Configure to emit an audible alarm when readings are outside limits
  • Programmable via USB interface using industry standard SCPI commands
These benchtop multimeters provide accurate and reliable measurements for a wide range of applications for use in the service & repair, manufacturing, educational, and test & measurement fields that require basic measurements. Meters offer a maximum measurement rate of up to 25 readings/sec that is selectable between slow, medium, and fast, and are an economical solution for users requiring a basic and dependable meter with a broad range of features.
$543.00 - $665.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
B&K Precision 2831E 4.5 Digit Digital Bench Multimeter EW-20036-39 Mfr # 2831E
COMPARE
220 0.03% AC DC Yes
$543.00USD / Each
B&K Precision 5491B 4.75 Digit Digital Bench Multimeter EW-20036-40 Mfr # 5491B
COMPARE
220 AC DC Yes
$665.00USD / Each
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat