Bacharach® Vacuum Pump
Rotary vane pumps designed for quick evacuation of refrigeration systems
  • High performance vacuum pump—choose from 1.7 CFM and 5.3 CFM models
  • Two-stage design—the second stage starts pumping at a lower pressure to pull a deeper ultimate vacuum
  • Anti-oil-return—check valve prevents pump-oil from entering the evacuated system after the pump stops
  • Environment friendly—built-in filter eliminates oil mist, and an oil-gas separator removes oil contamination from the exhaust
  • Aluminum alloy motor casing—motor utilizes strong light aluminum alloy, which guarantees cool, continuous operation
  • Compact design—motor and pump are integrated
  • Large starting torque—designed for low temperature and low voltage operation, the pump will start in temperatures ≥ 40°F (≥ 5°C), at its rated input voltage of 110 VAC ±10%
  • Oil circulation design—offset rotary vanes work in conjunction with the vacuum pump oil to create high compression within pumping chamber
  • Auto Power-off feature
The Model QV2 and QV5 are two-stage, rotary vane vacuum pumps designed to evacuate moisture and air from refrigeration systems.
$308.00 - $438.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Air Velocity Accuracy Air Velocity Resolution Max Air Velocity (ft/min)
Availability Pricing
Bacharach 2002-0001 Vacuum Pump, rotary vane, 1.7 cfm, 25 micron, 1/4 HP EW-48501-01 Mfr # 2002-0001
COMPARE
$308.00USD / Each
Bacharach 2002-0005 Vacuum Pump, rotary vane, 5.3 cfm, 25 micron, 1/2 HP EW-48501-02 Mfr # 2002-0005
COMPARE
$438.00USD / Each