Basic Slide Dryer
A convenient way to rapidly dry slides
  • Lexan® front for easy viewing of samples
  • Thermal switch prevents dryer from overheating
  • Accommodates up to two slide stainer racks
This slide dryer is designed to quickly dry slides in a temperature-controlled environment. Convection system circulates heated air over the surface of the slides. Access the drying chamber through a hinged lid at the top. The lid features an opening at the top that accepts any standard laboratory thermometer. The air temperature can be controlled between 30 to 70°C. Indicator light remains on when dryer is in use. The heater shuts off automatically in the event the slide dryer overheats. Holds up to two standard staining racks.
See our wide variety of glass thermometer offerings or call our Application Specialists to order.
$2,390.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
TBS SD-II-120-CP SD-II-120 Basic Slide Dryer, 30 to 70C; 120 VAC, 50/60 Hz EW-48577-51 Mfr # SD-II-120-CP
COMPARE
110 to 120 SD-II-120 Basic Slide Dryer, 30 to 70C; 120 VAC, 50/60 Hz
$2,390.00USD / Each