This product is now discontinued, View Related Products

Bathocuproine, 0.1% (w/v) Aqueous Solution

Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Ricca Chemical Company 905-16 Bathocuproine, 0.1% (w/v) Aqueous Solution (500 mL) EW-86970-05 Mfr # 905-16
COMPARE
Includes 52698-84-7 and 7732-18-5
Call For Price
Ricca Chemical Company 905-4 Bathocuproine, 0.1% (w/v) Aqueous Solution (120 mL) EW-86989-54 Mfr # 905-4
COMPARE
Includes 52698-84-7 and 7732-18-5
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat