Bausch & Lomb Hastings Measuring Magnifiers
Provides sharp, distinct images without distortion
  • Lens slides are coated black to block glare
  • Designed with precision optics and ergonomic properties for increased comfort during use
Optical precision comes from bonding together a double convex element and two concave elements of different types of glass. The transparent body allows illumination to fall on magnified area, the body is threaded so you can focus the lens.
Optional measuring scales (sold separately) screw onto the bottom of the lens to provide easy-to-see measurments.
$125.25 - $126.25USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Magnification Viewing Area Description
Availability Pricing
Bausch & Lomb Hastings Measuring Magnifier, 7X, 0.78" Diameter EW-03882-10 Mfr # 81-34-35
COMPARE
7x 0.78" (19.8 mm) diameter Hastings measuring magnifier; magnification; 7x
$125.25USD / Each
Bausch & Lomb Hastings Measuring Magnifier, 10X, 0.62" Diameter EW-03882-50 Mfr # 81-34-33
COMPARE
10x 0.62" (15.8 mm) diameter Hastings measuring magnifier; magnification; 10x
$126.25USD / Each