Bel-Art 96 Deep-Well Plates, Automation Compliant
Conforms to SBS/ANSI/SLAS footprints
  • DNase, RNase, and pyrogen free
  • Manufactured in an ISO Class 8 cleanroom
  • Alphanumeric marking and cut-angle mark for easy referencing
  • Can be stored at -80°C
  • Autoclavable up to 121°C
These 96 Deep-Well Plates can be easily used with 8- and 12-channel pipettes or with automation systems. They are made of virgin polypropylene. Applications include human genetic testing, forensic sciences, pathogen detection, food microbiology, veterinary science, and infectious disease testing. The plates come nonsterile.
$52.00USD / Pkg of 5
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sterile Well Shape Well Volume (mL)
Availability Pricing
Bel-Art 96 Deep-Well Plates, Automation Compliant, 2.2 mL, V-Bottom; 5/PK EW-07898-03 Mfr # 378600007
COMPARE
No V-bottom 2.2
$52.00USD / Pkg of 5
Bel-Art 96 Deep-Well Plates, Automation Compliant, 2.0 mL, U-Bottom; 5/PK EW-07898-04 Mfr # 378600008
COMPARE
No U-bottom 2
$52.00USD / Pkg of 5