Bel-Art Heavy-Duty Biohazard Disposal Bags and Stands
Indicator patch darkens when autoclaved
  • Heavy-duty bag resists tears and leaks
  • Autoclavable at 300°F (149°C)
These extra-strength, 2-mil thick bags are bright orange red. High-molecular weight polypropylene bags are printed with the biohazard warning symbol and precautionary procedures in four languages. The sterilization indicator patch darkens when autoclaved.
Optional bag stands are made of aluminum tubing with baked on epoxy coating. The stands fold flat for convenient storage and include a polystyrene safety drip tray that securely supports the bag in the stand. Order stands and plastic ties separately.
$253.00 - $701.00USD / Pkg of 200
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Gal) Length (in) Width (in)
Availability Pricing
Bel-Art Heavy-Duty Biohazard Disposal Bags, 14"W x 19"L EW-06495-00 Mfr # F13165-1419
COMPARE
4 19 14
$253.00USD / Pkg of 200
Bel-Art Heavy-Duty Biohazard Disposal Bags, 19"W x 23"L EW-06495-02 Mfr # F13165-1923
COMPARE
9 23 19
$427.00USD / Pkg of 200
Bel-Art Heavy-Duty Biohazard Disposal Bags, 25"W x 35"L EW-06495-04 Mfr # F13165-2535
COMPARE
12 35 25
$701.00USD / Pkg of 200