Quantek Instruments Benchtop Oxygen/Carbon Dioxide Analyzers
Ensure product quality and safety
  • Save time and money by preventing spoilage
Use this benchtop analyzer for continuous package testing in the laboratory or at the packing line. Analyzer features a heavy-duty, maintenance-free sensor that compensates for slight temperature variations. Unlike other sensors, accuracy is not affected by flow rate, heat, or hydrocarbon gases. To sample food packs, insert the needle tip of the probe into the food pack through a foam seal pad. Seal pad provides a tight seal and prevents air from leaking into the package through the puncture hole. Press the pump button to draw a sample from the package. The built-in pump draws in the precise amount of sample required for analysis, then turns off after a pre-set sampling time.
$3,993.00 - $5,244.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Gases Measured Description
Availability Pricing
COMPARE
O2 Benchtop Headspace OAnalyzer
$3,993.00USD / Each
Quantek Instruments 902D Benchtop Headspace O2/COAnalyzer EW-10360-70 Mfr # 902D
COMPARE
O2/CO2 Benchtop Headspace O2/COAnalyzer
$5,244.00USD / Each