This product is now discontinued, View Related Products

Fisher BioReagents β-Mercaptoethylamine Hydrochloride, 99+%

  • Assay - ≈98%
  • FTIR - Conforms to Standard
  • Melting Point - 58°–73°C
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical BP2664100 beta-Mercaptoethylamine Hydrochloride (100g) EW-88061-81 Mfr # BP2664100
COMPARE
156-57-0
Call For Price
Fisher Chemical BP266425 beta-Mercaptoethylamine Hydrochloride (25g) EW-88061-82 Mfr # BP266425
COMPARE
156-57-0
Call For Price
Fisher BioReagents BP26645 beta-Mercaptoethylamine Hydrochloride (5g) EW-88061-83 Mfr # BP26645
COMPARE
156-57-0
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat