This product is now discontinued, View Related Products
BetaGauge 330 Hand Held Pressure Calibrator 56998
Electric Pump Generates Pressure to 150 psi
  • The 330 comes in 3 ranges of 30 PSI, 150 PSI and 300 PSI. The 300 PSI version uses a manually operated handpump.
  • The compact, rugged design operates on eight (8) standard AA Alkaline batteries.
  • Performs approximately 300 calibration cycles to 150 PSI on one set of batteries.
The new BetaGauge 330 Pressure Calibrator provides a number of convenience features. Switch test can be performed on internal or external pressure input. Percent (%) Error and damping functions are provided. Up to five frequently used setups can be stored and retrieved with one button recall.
A Pt100 RTD input is provided for temperature measurements accurate to ±0.1 °C (0.2 °F).
In addition, the BetaGauge 330 measures and simulates 4-20 mA loop current signals. It can measure up to 30 VDC. An internal 24 VDC Instrument Power Supply supplies power to a transmitter under test.
4 PRODUCT OPTIONS
Compareitem specifications
Max Pressure Pressure Accuracy Pressure Resolution
Availability Pricing
Martel 1919793 Handheld Pressure Calibrator Kit; Electric/30 psi EW-68510-00 Mfr # 1919793
COMPARE
Call For Price
Martel 1919782 Handheld Pressure Calibrator Kit; Electric/150 psi EW-68510-01 Mfr # 1919782
COMPARE
Call For Price
Martel 1919790 Handheld Pressure Calibrator Kit; Manual/300 psi EW-68510-02 Mfr # 1919790
COMPARE
Call For Price
Martel BG330-ASSYKIT Accessory Kit for the 330 Calibrator EW-68510-03 Mfr # 1919801