Bev-A-Line IV Tubing
Applications: Safe for food, beverage, drug, lab, and general use—yet much more affordable than PTFE tubing. Safe for most alcohols; avoid use with strong solvents.
Characteristics: Polyethylene liner with ethyl vinyl acetate shell. No plasticizers. Nontoxic and nonhemolytic. Heat-bondable. Translucent.
Certification: Resin is FDA-compliant (liner: 21 CFR 177.1520; shell 21 CFR 177.1350). Meets USP Class VI, USDA, and National Formulary requirements.
Temperature range: –30 to 160°F (–51 to 71°C)
Sterilize: With ethylene oxide or dry heat.
$42.00 - $420.00USD / Pkg of 1
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
72 1/8 1/4
$42.00USD / Pkg of 1
Bev-A-Line B-13 IV Tubing, 1/4"ID x 3/8"OD, 50 ft/pack
Buy More Save More
EW-06490-15 Mfr # B-13
COMPARE
61 9/64 3/8
$66.00USD / Pkg of 1
Bev-A-Line B-19 IV Tubing, 3/8"ID x 1/2"OD, 50 ft/pack
Buy More Save More
EW-06490-17 Mfr # B-19
COMPARE
50 3/8 1/2
$95.50USD / Pkg of 1
Bev-A-Line B-25 IV Tubing, 1/2"ID x 5/8"OD, 50 ft/pack
Buy More Save More
EW-06490-19 Mfr # B-25
COMPARE
33 1/2 5/8
$128.00USD / Pkg of 1
Bev-A-Line B-27 IV Tubing, 50 psi at 70°F, 1/2" (12.8 mm) ID
Buy More Save More
EW-06490-20 Mfr # B-27
COMPARE
50 1/2 3/4
$257.00USD / Pkg of 1
Bev-A-Line B-38 IV Tubing, 3/4"ID x 1"OD, 50 ft/pack
Buy More Save More
EW-06490-39 Mfr # B-38
COMPARE
39 3/4 1
$354.00USD / Pkg of 1
Bev-A-Line B-45 IV Tubing, 1"ID x 1-1/4"OD, 50ft/pack
Buy More Save More
EW-06490-40 Mfr # B-45
COMPARE
28 1 1 1/4
$420.00USD / Pkg of 1
Showing 1- 7 of 7