Big Blue Cartridge Housings
Flow rates up to 50 GPM in a compact, relatively low-cost package
These systems offer up to four times the filtration area than standard cartridge systems, which makes them ideal for high-flow processes. A wide range of cartridges is offered for numerous applications. Order housings and cartridges separately.
Housing and cap are constructed of durable polypropylene and sealed with a Buna-N O-ring. Housing is suitable to 100°>F (37°>C); maximum operating pressure is 100 psi (6.9 bar).
$63.00 - $105.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Filter Length (in) Port Size Relief Valve
Availability Pricing
Big Blue 150239 Industrial filter housing; 10"L, 1-1/2" NPT(F), w/ relief valve EW-01507-76 Mfr # 150239
COMPARE
10 1.5" NPT(F) Yes
$71.50USD / Each
Big Blue 150468 Industrial filter housing; 20"L, 3/4" NPT(F) EW-01507-77 Mfr # 150468
COMPARE
20 3/4" NPT(F) No
$97.50USD / Each
Big Blue 150467 Industrial filter housing; 20"L, 3/4" NPT(F), w/ relief valve EW-01507-78 Mfr # 150467
COMPARE
20 3/4" NPT(F) Yes
$95.50USD / Each
Big Blue 150470 Industrial filter housing; 10"L, 3/4" NPT(F) EW-01508-65 Mfr # 150470
COMPARE
10 0.75" NPT(F) No
$68.00USD / Each
Big Blue 150469 Industrial filter housing; 10"L, 3/4" NPT(F), w/ relief valve EW-01508-67 Mfr # 150469
COMPARE
10 3/4" NPT(F) Yes
$71.00USD / Each
Big Blue 150238 Industrial filter housing; 10"L, 1" NPT(F) EW-01508-70 Mfr # 150238
COMPARE
10 1" NPT(F) No
$63.00USD / Each
Big Blue 150237 Industrial filter housing; 10"L, 1" NPT(F), w/ relief valve EW-01508-71 Mfr # 150237
COMPARE
10 1" NPT(F) Yes
$65.00USD / Each
Big Blue 150240 Industrial filter housing; 10"L, 1-1/2" NPT(F) EW-01508-75 Mfr # 150240
COMPARE
10 1.5" NPT(F) No
$71.50USD / Each
Big Blue 150234 Industrial filter housing; 20"L, 1" NPT(F) EW-29802-00 Mfr # 150234
COMPARE
20 1" NPT(F) No
$96.50USD / Each
Big Blue 150233 Industrial filter housing; 20"L, 1" NPT(F), w/ relief valve EW-29802-01 Mfr # 150233
COMPARE
20 1" NPT(F) Yes
$94.50USD / Each
Big Blue 150236 Industrial filter housing; 20"L, 1-1/2" NPT(F) EW-29802-10 Mfr # 150236
COMPARE
20 1.5" NPT(F) No
$102.00USD / Each
Big Blue 150235 Industrial filter housing; 20"L, 1-1/2" NPT(F), w/ relief valve EW-29802-11 Mfr # 150235
COMPARE
20 1.5" NPT(F) Yes
$105.00USD / Each
Showing 1- 12 of 12
Need Help with this product?
Call Us At1-847-549-7600
Live chat