This product is now discontinued, View Related Products
BIO-TEK ELx800 Universal Microplate Reader
Optional 72/96 Terasaki and 3840well capacity, UV 340nm capability
  • Stores up to 75 custom assay definitions and 8 test results
  • Extensive onboard data analysis and optional software which offers more flexible data manipulation
The Elx800 microplate readers feature on board data reduction and reporting. Reads 6-, 12-, 24--, 48-, and 96-well plate microplates. The model 13055-52 can read 384-well and 60-, 72- and 96-well Terasaki plates. Wavelength range from 400 to 750nm. Model 13055-54 optics to extend wavelength range down to 340nm.
Microplate readers feature a 2x24-character LCD and convenient soft-keys. Extensive curve fitting options include linear, cubic, quadratic, 4-p, log-logit, cubicspline, and point-to-point. Memory stores 25 standard curves and up to 8 microplate test results. Features auto or manual plate layout for blanks, controls, standards, and samples. Also offers control and assay validation, transformation and formula applications, and cutoff and call criteria.
Onboard software is easy to program to run sophisticated assays and analyzed without the need of computers. Additional software 13055-56 offers easy user interface with flexible data manipulation and curve fitting for basic endpoint, kinetic, multi-wavelength and spectral scan requirements. Software package includes software on CD, and three user licenses. What’s included: 340 (13055-54 only), 405, 450, 490, 630nm filters, dust cover, parallel printer cable (25-pin male and 36-pin male), serial cable (null modem, 9-pin female to 25-pin female), operator’s manual. Additional filters are available range from 304 to 750nm.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Material Sterile Surface Treatment
Availability Pricing
BioTek ELX800 Microplate reader, 400 to 750 nm, 6, 12, 24, 48, 96 well EW-13055-50 Mfr # ELX800
COMPARE
Call For Price
BioTek ELX800NB Microplate reader, 400 to 750 nm, 6, 12, 24, 48, 72/96, Terasaki, 96, 384 well EW-13055-52 Mfr # ELX800NB
COMPARE
Call For Price
BioTek ELX800UV Microplate reader, 340 to 750 nm, 6, 12, 24, 48, 96 well EW-13055-54 Mfr # ELX800UV