Biohazard Autoclave Gloves

Heat resistant to 450°F (232°C)
  • Heat resistant to 450°F (232°C)
Use these soft, pliant, and all-cotton terry cloth gloves to handle hot objects fresh from the autoclave or oven. Excellent for use in any environment where heat protection is required. Bright orange for increased visibility. Machine wash, tumble dry.
$48.00 - $63.00USD / Pair of 1

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size
Availability Pricing
Scienceware 13201 Biohazard Autoclave Gloves, 232C max, 13" L
Always in Stock
EW-06496-40 Mfr # 13201-0000
COMPARE
13 One size fits all
$48.00USD / Pair of 1
Scienceware H13201-0001 Biohazard Autoclave Gloves, 232C max, 18.5" L EW-06496-42 Mfr # H13201-0001
COMPARE
18 1/2 One size fits all
$63.00USD / Pair of 1
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat