This product is now discontinued, View Related Products

Bis-Acrylamide

  • Assay - ≥95%
  • pH of a 1% Solution in 0.1M NaCl - ≥5.0
  • Absorbance of a 1% Solution at 290nm - ≤0.2
  • Electrophoresis - To pass test
  • DNase - Not detected
  • RNase - Not detected
  • Protease - Not detected
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical BP171100 Bis-acrylamide (Electrophoresis) (100g) EW-88057-94 Mfr # BP171100
COMPARE
110-26-9
Call For Price
Fisher Chemical BP17125 Bis-acrylamide (Electrophoresis) (25g) EW-88057-95 Mfr # BP17125
COMPARE
110-26-9
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat