This product is now discontinued, View Related Products
BK Series 830C and 890C Capacitance Meters
  • 5 digit, 11000 count display
  • Automatic polarity indication
  • Ideal for measuring values, inspection, sorting, and testing capacitors against standard tolerances
  • Visible and audible tolerance mode assists in sorting capacitors
  • Min/max/average, data hold, with manual or auto trigger, and relative modes
  • Comparison mode with 25 sets of selectable high/low limits
  • Mini USB interface with included cable
  • Datalogging software available for download
  • AC power adapter (model 830C only)
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
B&K Precision 830C Capacitance tester with test leads EW-20042-53 Mfr # 830C
COMPARE
Call For Price
B&K Precision 890C Capacitance tester, autoranging, with test leads EW-20042-54 Mfr # 890C