Black Viton® FDA-Compliant Tubing
Great for high-temperature food and lab applications
  • Withstands high temperatures up to 204°C (400°F)
  • Excellent chemical resistance—great for transferring acids and solvents
  • Black tubing keep light sensitive samples safe
  • Soft and flexible—use with barbed fittings
  • Meets FDA 21CFR 177.2600 and USP Class VI
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Use this Viton® tubing in food or laboratory applications where FDA compliance is required. The tubing delivers excellent wear, abrasion, heat, UV, and ozone resistance, offering reliable performance every time. Viton tubing exhibits excellent chemical resistance, making it ideal for transferring acids and solvents. Each label includes the part number, batch number, cure date, length, and compound number for easy referencing.
$115.00 - $2,000.00USD / Pkg of 1
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Viton FDA Tubing, Black, 60D, 1/16" ID x 1/8" OD; 50 Ft
New Products
EW-50131-91 Mfr # T60062/0125A
COMPARE
1/16 1/8 1/32
$115.00USD / Pkg of 1
Viton FDA Tubing, Black, 60D, 1/8" ID x 3/16" OD; 50 Ft
New Products
EW-50131-92 Mfr # T60125/0187A
COMPARE
1/8 3/16 1/32
$145.00USD / Pkg of 1
Viton FDA Tubing, Black, 60D, 3/16" ID x 1/4" OD; 50 Ft
New Products
EW-50131-93 Mfr # T60187/0250A
COMPARE
3/16 1/4 1/32
$180.00USD / Pkg of 1
Viton FDA Tubing, Black, 60D, 1/4" ID x 5/16" OD; 50 Ft
New Products
EW-50131-94 Mfr # T60250/0312A
COMPARE
1/4 5/16 1/32
$210.00USD / Pkg of 1
Viton FDA Tubing, Black, 60D, 5/16" ID x 7/16" OD; 50 Ft
New Products
EW-50131-95 Mfr # T60312/0437A
COMPARE
5/16 7/16 1/16
$450.00USD / Pkg of 1
Viton FDA Tubing, Black, 60D, 3/8" ID x 1/2" OD; 50 Ft
New Products
EW-50131-96 Mfr # T60375/0500B
COMPARE
3/8 1/2 1/16
$530.00USD / Pkg of 1
Viton FDA Tubing, Black, 60D, 1/2" ID x 5/8" OD; 50 Ft
New Products
EW-50131-97 Mfr # T60500/0625B
COMPARE
1/2 5/8 1/16
$675.00USD / Pkg of 1
Viton FDA Tubing, Black, 60D, 5/8" ID x 3/4" OD; 50 Ft
New Products
EW-50131-98 Mfr # T60625/0750B
COMPARE
5/8 3/4 1/16
$830.00USD / Pkg of 1
Viton FDA Tubing, Black, 60D, 3/4" ID x 1" OD; 50 Ft
New Products
EW-50131-99 Mfr # T60750/1000B
COMPARE
3/4 1 1/8
$2,000.00USD / Pkg of 1
Showing 1- 9 of 9