Bovine Serum Albumin Solution
  • Total Protein (Bradford) - 18-22mg/mL
  • Optical Absorbance (dilution, 1:40 in water)
  • &tab;at 280nm - 0.25-0.35
  • Electrophoresis - To pass test
  • DNase - Not detected
  • RNase - Not detected
  • Protease - Not detected
$118.00 - $425.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher BioReagents BP6751 Bovine Serum Albumin Solution (20mg/ mL) (1 mL) EW-88068-33 Mfr # BP6751
COMPARE
9048-46-8 and 26628-22-8
$118.00USD / Each
Fisher BioReagents BP6755 Bovine Serum Albumin Solution (20mg/ mL) (5 mL) EW-88068-34 Mfr # BP6755
COMPARE
9048-46-8 and 26628-22-8
$425.00USD / Each