This product is now discontinued, View Related Products

Brady BMP 21 Laboratory Label Printer

Identify your lab samples quickly and legibly
  • One-handed operation with large display and backlight
  • Advanced serialization, barcoding and symbols
  • Cuts and holds printed labels until you need them
The BMP 21 portable label printer is an all-in-one labeler. With label materials designed for a variety of environments, the BMP 21 portable printer can quickly and easily create clear, legible labels that stay in place for years — despite extreme temperatures and curved or highly textured surfaces. Printer operates on either six AA batteries or AC power adapter (neither included; order separately).


Print labels in sizes specifically made for vials and slides. Vial size fonts are preprogrammed for 0.6, 1.5, and 2.0 mL. The laboratory-specific labels resist most chemicals, including alcohol, ethanol, and the solvent DMSO. Choose from 3/8", 1/2", and 3/4" wide labels in white or clear tape. Labels are designed for normal to extreme conditions, like liquid nitrogen or hot water baths.

Call For Price

10 variations of this product are available.

See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Brady BMP21 BMP 21 Laboratory Label Printer with 1 White Polylabel Cartridge EW-65533-12 Mfr # BMP21
COMPARE
BMP 21 Laboratory Label Printer with 1 White Polylabel Cartridge
Call For Price
Brady M21-375-430 BMP 21 Label Cartridge; 3/8"W, Poly, Clear EW-65533-14 Mfr # M21-375-430
COMPARE
BMP 21 Label Cartridge; 3/8\"W, Poly, Clear
Call For Price
Brady M21-375-580-WT BMP 21 Label Cartridge; 3/8"W, Permanent Vinyl, White EW-65533-16 Mfr # M21-375-580-WT
COMPARE
BMP 21 Label Cartridge; 3/8\"W, Permanent Vinyl, White
Call For Price
Brady M21-375-461 BMP 21 Label Cartridge; 3/8"W, Self-Laminating Poly, White EW-65533-18 Mfr # M21-375-461
COMPARE
BMP 21 Label Cartridge; 3/8\"W, Self-Laminating Poly, White
Call For Price
Brady M21-500-430 BMP 21 Label Cartridge; 1/2"W, High-Performance Poly, Clear EW-65533-20 Mfr # M21-500-430
COMPARE
BMP 21 Label Cartridge; 3/4\"W, High-Performance Poly, White
Call For Price
Brady M21-500-580-WT BMP 21 Label Cartridge; 1/2"W, Permanent Vinyl, White EW-65533-22 Mfr # M21-500-580-WT
COMPARE
BMP 21 Label Cartridge; 1/2\"W, Permanent Vinyl, White
Call For Price
Brady M21-500-461 BMP 21 Label Cartridge; 1/2"W, Self-Laminating Poly, White EW-65533-24 Mfr # M21-500-461
COMPARE
BMP 21 Label Cartridge; 1/2\"W, Self-Laminating Poly, White
Call For Price
Brady M21-750-461 BMP 21 Label Cartridge; 3/4"W, Self-Laminating Poly, White EW-65533-26 Mfr # M21-750-461
COMPARE
BMP 21 Label Cartridge; 3/4\"W, Self-Laminating Poly, White
Call For Price
Brady M21-750-488 BMP 21 Laboratory Label Printer Cartridge; 3/4" H, Poly, White EW-65533-28 Mfr # M21-750-488
COMPARE
BMP 21 Laboratory Label Printer Cartridge; 3/4\" H, Poly, White
Call For Price
Brady BMP21-AC BMP 21 Laboratory Label Printer AC Adapter; 120 VAC EW-65533-30 Mfr # M-AC-110937
COMPARE
BMP 21 Laboratory Label Printer AC Adapter; 120 VAC
Call For Price
Showing 1- 10 of 10

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat