BRAND® Disposable Fluorimetry Cuvettes
Four clear sides with 10 mm optical path to fit most standard fluorometers
  • Choose PS or special UV-transparent pastic with ultra-low auto-fluroescence
  • Mold cavity matched for consistency
BRAND® fluorimetry cuvettes are precision molded for best optical clarity and uniform thickness to optimize transmission while minimizing refraction. All cuvettes are packaged in low-dust, non-scratching, expanded polystyrene trays containing 100 mold-cavity matched cuvettes.
$27.50 - $119.00USD / Pkg of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Materials of Construction Min Wavelength (nm) Description
Availability Pricing
BRAND Macro Fluorimetry Cuvette, PS, 10 mm; 100/PK EW-06343-00 Mfr # 759030
COMPARE
Polystyrene 340 Fluorimetry Cuvette, PS, 10 mm; 100/PK
$27.50USD / Pkg of 100
BRAND Macro Fluorimetry Cuvette, PS, 10 mm; 500/PK EW-06343-02 Mfr # 759035
COMPARE
Polystyrene 340 Fluorimetry Cuvette, PS, 10 mm; 500/PK
$113.50USD / Pkg of 500
BRAND Macro Fluorimetry Cuvette, UV-Transparent Plastic, 10 mm; 100/PK EW-06343-04 Mfr # 759125
COMPARE
Polymer 230 Fluorimetry Cuvette, UV-Transparent Plastic, 10 mm; 100/PK
$119.00USD / Pkg of 100