BRAND® SAN Imhoff Sedimentation Cones

Easy drainage with bottom screw on cap
  • 1000 mL volume
  • Graduated per DIN 12672
  • Withstands temperatures up to 85° C
This settling cone is made of styrene acrylonitrile to offer exceptional optical clarity for measuring of settled solids.
$45.00 - $383.00USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bottle Type Material Sample Volume
Availability Pricing
BRAND 388000 Imhoff Sedimentation Cone, SAN, 1000 mL
Always in Stock
EW-06102-38 Mfr # 388000
COMPARE
Styrene Acrylonitrile Graduated to 1000 mL
$45.00USD / Each
BrandTech 388050 Rack for 2 1000 mL Imhoff Sedimentation Cones, PMMA; 1/EA EW-06102-39 Mfr # 388050
COMPARE
$383.00USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat