BRAND® Transferpette® Single-Channel Electronic Pipettors
Intuitive operation in an "Ergonomics Approved" pipettor
  • Eliminate user variability
  • Easy maintenance
  • Features most popular functions
These single-channel electronic pipettors feature a soft-touch pipetting key, contoured handle, adjustable finger rest, and improved tip cone to reduce the operational forces needed for use. The prestigious TUV testing service, an OSHA-accredited independent testing lab, has given these electronic pipettors the “Ergonomic Approved” certification.
Large LCD screen clearly displays all settings. Each function has a dedicated control to reduce inadvertent volume changes or tip ejection. Rechargeable battery provides over 4000 pipetting cycles on a singe charge. Units can be disassembled for cleaning and pipette shaft may be removed and autoclaved for decontamination or use in sterile applications. Easy Calibration™ system reduces the need for off-site calibration and simplifies conformance with ISO/GLP guidelines.
Operating modes:
Pipetting mode aspirates and discharges a preset volume.
Mixing mode repeatedly aspirates and discharges a preset volume and display shows number of completed cycles.
Reverse mode is designed for high viscosity, high vapor pressure, and low surface tension liquids.
Dispensing mode discharges a preset number of small equal aliquots from the total aspirated volume.
$630.00 - $673.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Autoclavable Max Volume (mL) Min Volume (mL)
Availability Pricing
BRAND Transferpette Electronic Pipettor, Single-Channel, 0.5-10 µL; 110 V Charger EW-25016-00 Mfr # 705319
COMPARE
Yes 0.01 0.0005
$673.00USD / Each
BRAND Transferpette Electronic Pipettor, Single-Channel, 2-20 µL; 110 V Charger EW-25016-02 Mfr # 705320
COMPARE
Yes 0.02 0.002
$673.00USD / Each
BRAND Transferpette Electronic Pipettor, Single-Channel, 10-200 µL; 110 V Charger EW-25016-04 Mfr # 705323
COMPARE
Yes 0.2 0.02
$673.00USD / Each
BRAND Transferpette Electronic Pipettor, Single-Channel, 50-1000 µL; 110 V Charger EW-25016-06 Mfr # 705326
COMPARE
Yes 1 0.05
$673.00USD / Each
BRAND Transferpette Electronic Pipettor, Single-Channel, 250-5000 µL; 110 V Charger EW-25016-08 Mfr # 705327
COMPARE
Yes 5 0.25
$673.00USD / Each
BRAND Transferpette Electronic Pipettor,Single-Channel, 0.5-10uL; no Charger EW-25016-10 Mfr # 705339
COMPARE
Yes 0.01 0.0005
$630.00USD / Each
BRAND Transferpette Electronic Pipettor, Single-Channel, 2-20uL; no Charger EW-25016-12 Mfr # 705340
COMPARE
Yes 0.02 0.002
$630.00USD / Each
BRAND Transferpette Electronic Pipettor,Single-Channel, 10-200uL; no Charger EW-25016-14 Mfr # 705343
COMPARE
Yes 0.2 0.02
$630.00USD / Each
BRAND Transferpette Electronic Pipettor,Single-Channel, 50-1000 µL; no Charger EW-25016-16 Mfr # 705346
COMPARE
Yes 1 0.05
$630.00USD / Each
BRAND Transferpette Electronic Pipettor, Single-Channel, 0.25-5 mL EW-25016-18 Mfr # 705347
COMPARE
Yes 5 0.25
$630.00USD / Each
Showing 1- 10 of 10