This product is now discontinued, View Related Products
BrandTech Dispensette Organic Bottletop Dispenser, Digital Volume Setting
Dispensette® Organic Dispensers Ideal for the dispensing of concentrated acids, fuming acids, peroxides, and difficult organic solvents — such as THF and methylene chloride*. Bright yellow markings let you easily identify these organic bottletop dispensers.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Division (mL) Max Volume (mL) Min Volume (mL)
Availability Pricing
BrandTech 4731331 Dispensette Organic Bottletop Dispenser, Digital; 0.5-5 mL EW-07878-38 Mfr # 4731331
COMPARE
0.02 5 0.5
Call For Price
BrandTech 4731341 Dispensette Organic Bottletop Dispenser, Digital; 1-10 mL EW-07878-44 Mfr # 4731341
COMPARE
0.05 10 1
Call For Price
BrandTech 4731351 Dispensette Organic Bottletop Dispenser, Digital; 2.5-25 mL EW-07878-46 Mfr # 4731351
COMPARE
0.1 25 2.5
Call For Price
BrandTech 4731361 Dispensette Organic Bottletop Dispenser, Digital; 5-50 mL EW-07878-48 Mfr # 4731361
COMPARE
0.2 50 5