Brass Female/Male Mini Manual Ball Valves
  • Full flow design ensures maximum operating efficiency
  • Blow-out proof stems
Mini brass ball valves are designed for use in confined and hard-to-reach areas and are ideal for use on water and air service lines.
$22.60 - $30.70USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Cv Inlet Port Outlet Port
Availability Pricing
Parker Hannifin MV608-2 1/8" NPT brass female/male mini manual ball valve EW-30533-20 Mfr # MV608-2
COMPARE
1.2 1/8" NPT(F) 1/8" NPT(M)
$22.60USD / Each
Parker Hannifin MV608-4 1/4" NPT brass female/male mini manual ball valve EW-30533-22 Mfr # MV608-4
COMPARE
5.8 1/4" NPT(F) 1/4" NPT(M)
$23.95USD / Each
Parker Hannifin MV608-6 3/8" NPT brass female/male mini manual ball valve EW-30533-24 Mfr # MV608-6
COMPARE
3.9 3/8" NPT(F) 3/8" NPT(M)
$26.00USD / Each
Parker Hannifin MV608-8 1/2" NPT brass female/male mini manual ball valve EW-30533-26 Mfr # MV608-8
COMPARE
5.6 1/2" NPT(F) 1/2" NPT(M)
$30.70USD / Each