This product is now discontinued, View Related Products

Brilliant Green, 1% (w/v) Aqueous Solution

High quality, competitive pricing, and rapid service
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Form Volume (mL)
Availability Pricing
Ricca Chemical Company 1155-16 Brilliant Green, 1% (w/v) Aqueous Solution (500 mL) EW-86970-17 Mfr # 1155-16
COMPARE
Includes 633-03-4 and 7732-18-5 Liquid 500
Call For Price
Ricca Chemical Company 1155-4 Brilliant Green, 1% (w/v) Aqueous Solution (120 mL) EW-86989-66 Mfr # 1155-4
COMPARE
Includes 633-03-4 and 7732-18-5 Liquid 120
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat