British Standard Pipe Tapered Threaded (BSPT) Adapters
Convert NPT-threaded systems to BSP threads—or visa versa—with these stainless steel fittings.
Use BSP taper (BSPT) fittings for connections where pressure-tight joints are made on the thread, BSP parallel (BSPP) fittings where pressure-tight joints are not made on the thread (as in connections with peripheral seals). Stainless steel fittings are chemically resistant.
$23.50 - $101.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
Parker Hannifin Threaded adapter, stainless steel, 1/8" female NPT to 1/8" male BSPT
Buy More Save More
EW-31830-40 Mfr # 2-2K RA-SS
COMPARE
Stainless Steel
$23.50USD / Each
Parker Hannifin Threaded adapter, stainless steel, 1/4" female NPT to 1/4" male BSPT
Buy More Save More
EW-31830-41 Mfr # 4-4K RA-SS
COMPARE
Stainless Steel
$35.50USD / Each
Parker Hannifin Threaded adapter, stainless steel, 3/8" male BSPT to 3/8" female NPT
Buy More Save More
EW-31830-42 Mfr # 6-6K RA-SS
COMPARE
Stainless Steel
$62.00USD / Each
Parker Hannifin Threaded adapter, stainless steel, 1/2" female NPT to 1/2" male BSPT
Buy More Save More
EW-31830-43 Mfr # 8-8K RA-SS
COMPARE
Stainless Steel
$80.90USD / Each
Parker Hannifin 12-12K RA-SS Threaded adapter, stainless steel, 3/4" female NPT to 3/4" male BSPT
Buy More Save More
EW-31830-44 Mfr # 12-12K RA-SS
COMPARE
Stainless Steel
$87.50USD / Each
Parker Hannifin 2K-2 RA-SS Threaded adapter, stainless steel, 1/8" female BSPT to 1/8" male NPT
Buy More Save More
EW-31830-60 Mfr # 2K-2 RA-SS
COMPARE
Stainless Steel
$62.50USD / Each
Parker Hannifin Threaded adapter, stainless steel, 1/4" female BSPT to 1/4" male NPT
Buy More Save More
EW-31830-61 Mfr # 4K-4 RA-SS
COMPARE
Stainless Steel
$67.50USD / Each
Parker Hannifin Threaded adapter, stainless steel, 3/8" female BSPT to 3/8" male NPT
Buy More Save More
EW-31830-62 Mfr # 6K-6 RA-SS
COMPARE
Stainless Steel
$73.00USD / Each
Parker Hannifin Threaded adapter, stainless steel, 1/2" female BSPT to 1/2" male NPT
Buy More Save More
EW-31830-63 Mfr # 8K-8 RA-SS
COMPARE
Stainless Steel
$101.00USD / Each
Parker Hannifin Threaded adapter, stainless steel, 3/4" female BSPT to 3/4" male NPT
Buy More Save More
EW-31830-64 Mfr # 12K-12 RA-SS
COMPARE
Stainless Steel
$94.00USD / Each
Showing 1- 10 of 10