This product is now discontinued, View Related Products

Ricca Chemical Bromocresol Green-Methyl Red TS

High quality, competitive pricing, and rapid service
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Volume (mL)
Availability Pricing
Ricca Chemical Company 1221-16 Bromocresol Green-Methyl Red TS (500 mL) EW-86970-25 Mfr # 1221-16
COMPARE
Includes 76-60-8, 64-17-5, 67-56-1, 63451-28-5 and 7732-18-5 Components all ACS or USP or NF 500
Call For Price
Ricca Chemical Company 1221-4 Bromocresol Green-Methyl Red TS (120 mL) EW-86989-73 Mfr # 1221-4
COMPARE
Includes 76-60-8, 64-17-5, 67-56-1, 63451-28-5 and 7732-18-5 Components all ACS or USP or NF 120
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat