This product is now discontinued, View Related Products
Brookfield Rotational Viscometers
Economical digital viscometers with direct readout in centipoise
  • 18 speeds provide wide measuring capability
Measure viscosity in a wide range of centipoise (cp) for a variety of industrial and laboratory applications. The four-digit LCD ensures easy reading of both centipoise (cp) or millpascal seconds (mPaS), as well as percent torque, and speed. Display also shows spindle number.
Just flip a switch and turn a dial to set the speed and spindle code. Each model has 18 selectable rotational speeds, plus a set of standard spindles. Adjust rotational speed and spindle type to measure the desired centipoise range.
Order replacement spindles, optional temperature control units, small sample adapters, and other accessories separately.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cup Number Max Viscosity Range (cP) Power (VAC)
Availability Pricing
Brookfield AMETEK LVDVE115 Low-range viscometer, 115 VAC EW-98945-00 Mfr # LVDVE115
COMPARE
2000000 115
Call For Price
Brookfield AMETEK LVDVE230 Low-range viscometer, 230 VAC EW-98945-05 Mfr # LVDVE230
COMPARE
2000000 230
Call For Price
Brookfield AMETEK RVDVE115 Medium-range viscometer, 115 VAC EW-98945-10 Mfr # RVDVE115
COMPARE
13000000 115
Call For Price
Brookfield AMETEK RVDVE230 Medium-range viscometer, 230 VAC EW-98945-15 Mfr # RVDVE230
COMPARE
13000000 230
Call For Price
Brookfield AMETEK HBDVE115 High-range viscometer, 115 VAC EW-98945-20 Mfr # HBDVE115
COMPARE
104000000 115
Call For Price
Brookfield AMETEK HBDVE230 High-range viscometer, 230 VAC EW-98945-25 Mfr # HBDVE230
COMPARE
104000000 230