This product is now discontinued, View Related Products
BTX Electro Cell Fusion Systems
Exponential decay, square wave, and electro cell manipulatin systems
  • Available for US Sale only
Electro Cell Fusion Systems are versatile electro cell manipulation instruments. They generate a proprietary AC waveform for benign dielectrophoretic alignment of cells. With only microsecond switch-over time from AC to DC, efficient fusion can take place. After fusion, AC reapplication keeps cells together for the rounding off process. The DC square pulse section can be used as a stand alone powerful electroporation system with a wide range of voltages and pulse lengths capable of operating into low impedance loads.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
High Voltage Mode - Capacitance High Voltage Mode - Pulse Length High Voltage Mode - Voltage
Availability Pricing
BTX Embryo Manipulation Electro Cell Fusion System; 100-240 VAC EW-02894-00 Mfr # 45-0012
COMPARE
DC: 1 to 99 msec 30 to 3000
Call For Price
BTX Electro Cell Fusion System; 100-240 VAC EW-02894-02 Mfr # 45-0010
COMPARE
DC: 1 to 99 msec 30 to 3000
Call For Price
BTX Electroporation Electro Cell Fusion System; 100-240 VAC EW-02894-04 Mfr # 45-0011
COMPARE
DC: 1 to 99 msec 30 to 3000