EW-23123-61 Mfr # 008750
COMPARE 50 mL Uncoated 50 mL evaporator flask for Rotary Evaporators, 24/40 joint
2312361 EW-23123-62 Mfr # 008751
COMPARE 100 mL Uncoated 100 mL evaporator flask for Rotary Evaporators, 24/40 joint
2312362 EW-23123-63 Mfr # 008754
COMPARE 250 mL Uncoated 250 mL evaporator flask for Rotary Evaporators, 24/40 joint
2312363 EW-23123-64 Mfr # 008758
COMPARE 500 mL 500 mL evaporator flask for Rotary Evaporators, 24/40 joint
2312364 EW-23123-65 Mfr # 000440
COMPARE 1000 mL Uncoated 1000 mL evaporator flask for Rotary Evaporators, 24/40 joint
2312365 EW-23123-66 Mfr # 008765
COMPARE 2000 mL Uncoated 2000 mL evaporator flask for Rotary Evaporators, 24/40 joint
2312366 EW-23123-67 Mfr # 008767
COMPARE 3000 mL 3000 mL evaporator flask for Rotary Evaporators, 24/40 joint
2312367 EW-23123-68 Mfr # 047990
COMPARE 4000 mL 4000 mL evaporator flask for Rotary Evaporators, 24/40 joint
2312368 EW-23123-69 Mfr # 025261
COMPARE 500 mL Safety-coated 500 mL evaporator flask with safety-coating for Rotary Evaporators, 24/40 joint
2312369 EW-23123-70 Mfr # 020730
COMPARE 1000 mL Safety-coated 1000 mL evaporator flask with safety-coating 24/40 joint
2312370 EW-23123-71 Mfr # 025262
COMPARE 2000 mL Safety-coated 2000 mL evaporating flask with safety-coating 24/40 joint
2312371 EW-23123-72 Mfr # 025263
COMPARE 3000 mL Safety-coated 3000 mL evaporating flask with safety-coating 24/40 joint
2312372 EW-23123-73 Mfr # 047992
COMPARE 4000 mL Safety-coated 4000 mL evaporating flask with safety-coating 24/40 joint
2312373 EW-23123-74 Mfr # 011579
COMPARE 500 mL Uncoated 500 mL drying flask for powders, 24/40 joint
2312374 EW-23123-75 Mfr # 000420
COMPARE 1000 mL Uncoated 1000 mL drying flask for powders, 24/40 joint
2312375 EW-23123-76 Mfr # 011580
COMPARE 2000 mL Uncoated 2000 mL drying flask for powders, 24/40 joint
2312376 EW-23123-77 Mfr # 000421
COMPARE 50 mL Uncoated 50 mL receiving flask for Rotary Evaporators
2312377 EW-23123-78 Mfr # 000422
COMPARE 100 mL Uncoated 100 mL receiving flask for Rotary Evaporators
2312378 EW-23123-79 Mfr # 000423
COMPARE 250 mL Uncoated 250 mL receiving flask for Rotary Evaporators
2312379 EW-23123-80 Mfr # 000424
COMPARE 500 mL 500 mL receiving flask for Rotary Evaporators
2312380