Bud Industries Cable Glands
Ideal for Bud Industries NEMA enclosures
  • UL94V-2 flammability rating
  • -40 to 100°C operating temperature range
The cable glands consists of a black molded nylon body, hexagonal nut, cable nut, a rubber gasket, and cable seal. Cable seal maintains NEMA rating for cable entry or exit.
$3.40USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Bud Industries NG-9514 Cable Gland, PG-13.5 Mount Thread, 6 to 11 mm EW-94501-44 Mfr # NG-9514
COMPARE
Cable Gland, mounting size PG-13.5
$3.40USD / Each