Bud Industries TR-Series Enclosure for DIN Mount Controllers
Convert your DIN controllers into a portable desktop instrument
  • Corner gussets provides additional strength
  • Carrying handle may be used as tilting bail
The versitality makes it ideal to use as a portable enclosure, a desk top instrument case, or directly into a larger system. Front panel is contructed of 0.09" aluminum and can be easily stamped out to fit controllers and panel meters from 1/16 to 1/2 DIN sizes. Top and bottom panels are constructed from 0.05" aluminum and are easily removed for access to the dust-free interior. Enclosure is finished with a royal blue texture with a white frame. Enclosure comes completely assembled with all hardware included.
$297.00 - $340.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Outside Depth (in) Outside Height (in) Outside Width (in)
Availability Pricing
Bud Industries TR-6101 Enclosure for DIN-Mount Controllers, Medium Size EW-94503-18 Mfr # TR-6101
COMPARE
13.25 5.25 8.31
$297.00USD / Each
Bud Industries TR-6102 Enclosure for DIN-Mount Controllers, Medium/Lg Size EW-94503-20 Mfr # TR-6102
COMPARE
13.25 7 11.06
$340.00USD / Each
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat