Burkert Paddle-Wheel Sensors with Integral Fittings
  • Can be used with various liquids
These paddle-wheel sensors are ideal for measuring both neutral and aggressive, solid-free liquids. Built-in pipe fittings ensure simple installation into pipes from 1/2" up to 2". Mount sensor/fittings horizontally or vertically.
Choose from polyvinyl chloride (PVC), brass, or stainless steel (SS) sensor/fittings. PVC models come with true union fittings (solvent/fusion connection). Order brass and SS models with NPT(F) fittings for high temperature or high pressure applications. SS models are especially ideal for measuring aggressive fluids. Wetted parts are polyvinylidene fluoride, ceramic, fluoropolymer, and PVC, brass, or SS.
$539.00 - $1,570.00USD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Material Fitting Pipe Size Process Connection
Availability Pricing
Burkert 423950W Paddle Wheel Sensor w/ Fitting, PVC, 235 psi, 1/2" (DN15), true union EW-33117-00 Mfr # 423950W
COMPARE
PVC 1/2" (DN15) True union
$634.00USD / Each
Burkert 423986F Paddle Wheel Sensor w/ Fitting, brass, 350 psi, 1/2" (DN15), 1/2" NPT(F) EW-33117-05 Mfr # 423986F
COMPARE
Brass 1/2" (DN15) 1/2" NPT(F)
$539.00USD / Each
Burkert 424010L Paddle Wheel Sensor w/ Fitting, 316L SS, 350 psi, 1/2" (DN15), 1/2" NPT(F) EW-33117-10 Mfr # 424010L
COMPARE
316L SS 1/2" (DN15) 1/2" NPT(F)
$1,050.00USD / Each
Burkert 423951K Paddle Wheel Sensor w/ Fitting, PVC, 235 psi, 3/4" (DN20), true union EW-33117-15 Mfr # 423951K
COMPARE
PVC 3/4" (DN20) True union
$584.00USD / Each
Burkert 423 987 G Paddle Wheel Sensor w/ Fitting, brass, 350 psi, 3/4" (DN20), 3/4" NPT(F) EW-33117-20 Mfr # 423 987 G
COMPARE
Brass 3/4" (DN20) 3/4" NPT(F)
$592.00USD / Each
Burkert 424011H Paddle Wheel Sensor w/ Fitting, 316L SS, 350 psi, 3/4" (DN20), 3/4" NPT(F) EW-33117-25 Mfr # 424011H
COMPARE
316L SS 3/4" (DN20) 3/4" NPT(F)
$1,100.00USD / Each
Burkert 423952L Paddle Wheel Sensor w/ Fitting, PVC, 235 psi, 1" (DN25), true union EW-33117-30 Mfr # 423952L
COMPARE
PVC 1" (DN25) True union
$595.00USD / Each
Burkert 423988R Paddle Wheel Sensor w/ Fitting, brass, 350 psi, 1" (DN25), 1" NPT(F) EW-33117-35 Mfr # 423988R
COMPARE
Brass 1" (DN25) 1" NPT(F)
$610.00USD / Each
Burkert 424012A Paddle Wheel Sensor w/ Fitting, 316L SS, 350 psi, 1" (DN25), 1" NPT(F) EW-33117-40 Mfr # 424012A
COMPARE
316L SS 1" (DN25) 1" NPT(F)
$1,170.00USD / Each
Burkert 424013B Paddle Wheel Sensor w/ Fitting, 316L SS, 350 psi, 1-1/4" (DN32), 1-1/4" NPT(F) EW-33117-55 Mfr # 424013B
COMPARE
316L SS 1-1/4" (DN32) 1-1/4" NPT(F)
$1,310.00USD / Each
Burkert 423954N Paddle Wheel Sensor w/ Fitting, PVC, 235 psi, 1-1/2" (DN40), true union EW-33117-60 Mfr # 423954N
COMPARE
PVC 1-1/2" (DN40) True union
$684.00USD / Each
Burkert 423955P Paddle Wheel Sensor w/ Fitting, PVC, 235 psi, 2" (DN65), true union EW-33117-75 Mfr # 423955P
COMPARE
PVC 2" (DN65) True union
$708.00USD / Each
Burkert 423991C Paddle Wheel Sensor w/ Fitting, brass, 350 psi, 2" (DN65), 2" NPT(F) EW-33117-80 Mfr # 423991C
COMPARE
Brass 2" (DN65) 2" NPT(F)
$742.00USD / Each
Burkert 424015D Paddle Wheel Sensor w/ Fitting, 316L SS, 350 psi, 2" (DN65), 2" NPT(F) EW-33117-85 Mfr # 424015D
COMPARE
316L SS 2" (DN65) 2" NPT(F)
$1,570.00USD / Each
Showing 1- 14 of 14
Need Help with this product?
Call Us At1-847-549-7600
Live chat