Burkert Type 3233 316L Stainless Steel Diaphragm Valve
Fluids are hermetically sealed from operating system
  • Zero dead volume
  • Autoclavable
Choose from EPDM or PTFE wetted materials and sanitary or weld end connections. These compact, manually-operated diaphragm valves consist of a flow-optimized stainless steel body, diaphragm, and a manual actuator. The diaphragm is both a switch element and a sealing element and can be easily replaced. The valves have no dead volume and can be mounted to be self-draining — this enables high flow capacities and a variety of applications to be realized. The flow can be continually adjusted with the handwheel, making these valves extremely useful in applications in the food industry, biotechnology, and pharmaceutical markets.
$375.00 - $2,640.00USD / Each
27 variations of this product are available.
See All Product Options Below
27 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Outlet Port Wetted Materials
Availability Pricing
Burkert 98126050 (D051) 3233 316LSS 2-Way Diaphragm Valve, EPDM; 1/2"Tri-Clamp, 145psi EW-98516-28 Mfr # 292027
COMPARE
1/2" Tri-clamp 1/2" Tri-clamp 316L stainless steel, EPDM
$687.00USD / Each
Burkert 445873 3233 316LSS 2-Way Diaphragm Valve, EPDM; 3/4"Tri-Clamp, 150psi EW-98516-32 Mfr # 321142
COMPARE
3/4" Tri-clamp 3/4" Tri-clamp 316L stainless steel, EPDM
$1,212.00USD / Each
Burkert 445888 3233 316LSS 2-Way Diaphragm Valve, EPDM; 2"Tri-Clamp, 150psi EW-98516-38 Mfr # 445888
COMPARE
2" Tri-clamp 2" Tri-clamp 316L stainless steel, EPDM
$1,760.00USD / Each
Burkert 447980 (D051) 3233 316LSS 2-Way Diaphragm Valve, EPDM; 3/8" Weld-End, 145psi EW-98516-42 Mfr # 447980 (D051)
COMPARE
3/8" Weld end 3/8" Weld end 316L stainless steel, EPDM
$375.00USD / Each
Burkert 205977 (D050 + N017) 3233 316LSS 2-Way Diaphragm Valve, EPDM; 1/2" Weld-End, 145psi EW-98516-44 Mfr # 205977 (D050 + N017)
COMPARE
1/2" Weld end 1/2" Weld end 316L stainless steel, EPDM
$396.00USD / Each
Burkert 445573 3233 316LSS 2-Way Diaphragm Valve, EPDM; 1/2" Weld-End, 150psi EW-98516-46 Mfr # 445573
COMPARE
1/2" Weld end 1/2" Weld end 316L stainless steel, EPDM
$652.00USD / Each
Burkert 446545 (D051) 3233 316LSS 2-Way Diaphragm Valve, PTFE; 1/4"Tri-Clamp, 145psi EW-98516-56 Mfr # 446545 (D051)
COMPARE
1/4" Tri-clamp 1/4" Tri-clamp 316L stainless steel, PTFE
$696.00USD / Each
Burkert 446550 (D051) 3233 316LSS 2-Way Diaphragm Valve, PTFE; 3/8"Tri-Clamp, 145psi EW-98516-58 Mfr # 446550 (D051)
COMPARE
3/8" Tri-clamp 3/8" Tri-clamp 316L stainless steel, PTFE
$696.00USD / Each
Burkert 553269 (D051) 3233 316LSS 2-Way Diaphragm Valve, PTFE; 1/2"Tri-Clamp, 145psi EW-98516-60 Mfr # 553269 (D051)
COMPARE
1/2" Tri-clamp 1/2" Tri-clamp 316L stainless steel, PTFE
$633.00USD / Each
Burkert 445833 3233 316LSS 2-Way Diaphragm Valve, PTFE; 1/2"Tri-Clamp, 150psi EW-98516-62 Mfr # 445833
COMPARE
1/2" Tri-clamp 1/2" Tri-clamp 316L stainless steel, PTFE
$887.00USD / Each
Burkert 445838 3233 316LSS 2-Way Diaphragm Valve, PTFE; 3/4"Tri-Clamp, 150psi EW-98516-64 Mfr # 445838
COMPARE
3/4" Tri-clamp 3/4" Tri-clamp 316L stainless steel, PTFE
$1,218.00USD / Each
Burkert 445843 3233 316LSS 2-Way Diaphragm Valve, PTFE; 1"Tri-Clamp, 150psi EW-98516-66 Mfr # 445843
COMPARE
1" Tri-clamp 1" Tri-clamp 316L stainless steel, PTFE
$1,428.00USD / Each
Burkert 445848 3233 316LSS 2-Way Diaphragm Valve, PTFE; 1.5"Tri-Clamp, 150psi EW-98516-68 Mfr # 445848
COMPARE
1-1/2" Tri-clamp 1-1/2" Tri-clamp 316L stainless steel, PTFE
$1,857.00USD / Each
Burkert 445853 3233 316LSS 2-Way Diaphragm Valve, PTFE; 2"Tri-Clamp, 150psi EW-98516-70 Mfr # 445853
COMPARE
2" Tri-clamp 2" Tri-clamp 316L stainless steel, PTFE
$2,640.00USD / Each
Burkert 558789 (D051) 3233 316LSS 2-Way Diaphragm Valve, PTFE; 1/2" Weld-End, 145psi EW-98516-76 Mfr # 558789 (D051)
COMPARE
1/2" Weld end 1/2" Weld end 316L stainless steel, PTFE
$541.00USD / Each
Burkert 445608 3233 316LSS 2-Way Diaphragm Valve, PTFE; 1/2" Weld-End, 150psi EW-98516-78 Mfr # 445608
COMPARE
1/2" Weld end 1/2" Weld end 316L stainless steel, PTFE
$1,047.00USD / Each
Burkert 445613 3233 316LSS 2-Way Diaphragm Valve, PTFE; 3/4" Weld-End, 150psi EW-98516-80 Mfr # 445613
COMPARE
3/4" Weld end 3/4" Weld end 316L stainless steel, PTFE
$1,099.00USD / Each
Burkert 445618 3233 316LSS 2-Way Diaphragm Valve, PTFE; 1" Weld-End, 150psi EW-98516-82 Mfr # 445618
COMPARE
1" Weld end 1" Weld end 316L stainless steel, PTFE
$855.00USD / Each
Burkert 445623 3233 316LSS 2-Way Diaphragm Valve, PTFE; 1.5" Weld-End, 150psi EW-98516-84 Mfr # 445623
COMPARE
1-1/2" Weld end 1-1/2" Weld end 316L stainless steel, PTFE
$1,751.00USD / Each
Burkert 445628 3233 316LSS 2-Way Diaphragm Valve, PTFE; 2" Weld-End, 150psi EW-98516-86 Mfr # 445628
COMPARE
2" Weld end 2" Weld end 316L stainless steel, PTFE
$2,230.00USD / Each
Showing 1 - 20 of 27