Cadence Luer Adapter Fittings
Excellent durability
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured in an ISO 14644-1 Level 8 (formerly Class 100,000) clean room
  • Full material traceability on all items
$23.00 - $39.75USD / Each
23 variations of this product are available.
See All Product Options Below
23 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size-Port 2 (in) Connection Type-Port 1 Connection Type-Port 2
Availability Pricing
Cadence Luer Fitting, nickel-plated brass, female luer lock to male luer slip; 1/ea EW-41507-50 Mfr # 6170IND
COMPARE
Female luer Male luer slip
$23.25USD / Each
COMPARE
Female luer
$32.75USD / Each
COMPARE
Male luer slip
$31.50USD / Each
Cadence Luer Fitting, nickel-plated brass, male luer lock to male luer lock; 1/ea EW-41507-55 Mfr # 6177IND
COMPARE
Male luer lock Male luer lock
$38.00USD / Each
Cadence Luer Fitting, nickel-plated brass, female luer lock to female luer lock; 1/ea EW-41507-56 Mfr # 6182IND
COMPARE
Female luer Female luer
$25.75USD / Each
Cadence Luer Fitting, nickel-plated brass, male luer slip to male luer slip; 1/ea EW-41507-57 Mfr # 6183IND
COMPARE
Male luer slip Male luer slip
$28.75USD / Each
Cadence Luer Fitting Adapter, nickel-plated brass, male luer slip to 1/4" barb; 1/ea EW-41507-60 Mfr # 6152IND
COMPARE
1/4 Male luer slip
$23.00USD / Each
Cadence Luer Fitting Adapter, nickel-plated brass, male luer lock to 1/8" barb; 1/ea EW-41507-61 Mfr # 6156IND
COMPARE
1/8 Male luer lock
$28.50USD / Each
Cadence Luer Fitting Adapter, nickel-plated brass, male luer lock to 1/16" barb; 1/ea EW-41507-62 Mfr # 6157IND
COMPARE
5/8 Male luer slip
$28.50USD / Each
Cadence Luer Fitting Adapter, nickel-plated brass, female luer lock to 1/8" barb; 1/ea EW-41507-63 Mfr # 6158IND
COMPARE
1/8 Female luer
$24.00USD / Each
Cadence Luer Fitting Adapter, nickel-plated brass, female luer lock to 1/16" barb; 1/ea EW-41507-64 Mfr # 6159IND
COMPARE
5/8 Female luer
$30.00USD / Each
Cadence Luer Fitting Adapter, nickel-plated brass, female luer lock to 3/16" barb; 1/ea EW-41507-65 Mfr # 6168IND
COMPARE
3/16 Female luer
$23.25USD / Each
Cadence Luer Fitting Adapter, nickel-plated brass, male luer lock to 1/4" barb; 1/ea
Always in Stock
EW-41507-66 Mfr # 6169IND
COMPARE
1/4 Male luer lock
$23.75USD / Each
Cadence Luer Fitting Adapter, nickel-plated brass, male luer lock to 10-32 UNF; 1/ea EW-41507-70 Mfr # 6211IND
COMPARE
Male luer lock
$33.25USD / Each
Cadence Luer Fitting Adapter, nickel-plated brass, female luer lock to 10-32 UNF; 1/ea EW-41507-71 Mfr # 6212IND
COMPARE
Female luer
$35.00USD / Each
Cadence Luer Fitting Adapter, nickel-plated brass, male luer lock to 1/4-28 UNF; 1/ea EW-41507-73 Mfr # 6319IND
COMPARE
Male luer lock
$31.00USD / Each
Cadence Luer Fitting Adapter, nickel-plated brass, male luer lock to 1/8" NPT(M); 1/ea EW-41507-84 Mfr # 6330IND
COMPARE
Male luer lock
$34.50USD / Each
Cadence Luer Fitting Adapter, nickel-plated brass, male luer lock to 1/4" NPT(M); 1/ea EW-41507-85 Mfr # 6331IND
COMPARE
Male luer lock
$37.50USD / Each
Cadence Luer Fitting Adapter, nickel-plated brass, female luer lock to 1/8" NPT(M); 1/ea EW-41507-86 Mfr # 6332IND
COMPARE
Female luer
$38.50USD / Each
Cadence Luer Fitting Adapter, nickel-plated brass, female luer lock to 1/4" NPT(M); 1/ea EW-41507-87 Mfr # 6333IND
COMPARE
Female luer
$39.75USD / Each
Showing 1 - 20 of 23