Cadence Science Nickel-Plated Brass Luer Stopcocks
Exceptional durability
  • Easy to disassemble for cleaning
  • Stopcock handles turn 180°
  • Autoclavable
These Cadence Science luer stopcocks are constructed of nickel-plated brass for enhanced durability. These stopcocks offer worry-free cleaning with easy disassembly to save valuable time.
$131.00 - $486.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description Flow Pattern
Availability Pricing
Cadence Luer Stopcock Fitting, Nickel-Plated Brass, One-Way, Female Luer x Male Slip EW-41507-00 Mfr # 6019IND
COMPARE
Luer Stopcock Fitting, Nickel-Plated Brass, One-Way, Female Luer x Male Slip; 1/Ea One-Way
$131.00USD / Each
Cadence Luer Stopcock Fitting, Nickel-Plated Brass, One-Way, Female Luer x Male Luer Lock
Always in Stock
EW-41507-01 Mfr # 6021IND
COMPARE
Luer Stopcock Fitting, Nickel-Plated Brass, One-Way, Female Luer x Male Luer Lock; 1/Ea One-Way
$178.00USD / Each
Cadence Luer Stopcock Fitting, Nickel-Plated Brass, One-Way, Male Luer Lock x Male Luer Lock EW-41507-02 Mfr # 6023IND
COMPARE
Luer Stopcock Fitting, Nickel-Plated Brass, One-Way, Male Luer Lock x Male Luer Lock; 1/Ea One-Way
$172.00USD / Each
Cadence Luer Stopcock Fitting, Nickel-Plated Brass, Three-Way, Female Luer x Male Luer Lock x Female Luer EW-41507-10 Mfr # 6001IND
COMPARE
Luer Stopcock Fitting, Nickel-Plated Brass, Three-Way, Female Luer x Male Luer Lock x Female Luer; 1/Ea Three-Way
$205.00USD / Each
Cadence Luer Stopcock, Nickel-Plated Brass, Three-Way, Female Luer x Male Luer Lock x 3.5 mm ID EW-41507-11 Mfr # 6010IND
COMPARE
Luer Stopcock Fitting, Nickel-Plated Brass, Three-Way, Female Luer x Male Luer Lock x 3.5 mm Hose Barb; 1/Ea Three-Way
$192.00USD / Each
Cadence Luer Stopcock, Nickel-Plated Brass, Three-Way, Female Luer x Male Luer Lock x Side-Arm Female Luer EW-41507-12 Mfr # 6011IND
COMPARE
Luer Stopcock Fitting, Nickel-Plated Brass, Three-Way, Female Luer x Male Luer Lock x Side-Arm Female Luer; 1/Ea Three-Way
$171.00USD / Each
Cadence Luer Stopcock Fitting, Nickel-Plated Brass, Three-Way, Male Luer Lock x Male Luer Lock x Side-Arm Female Luer EW-41507-13 Mfr # 6014IND
COMPARE
Luer Stopcock Fitting, Nickel-Plated Brass, Three-Way, Male Luer Lock x Male Luer Lock x Side-Arm Female Luer; 1/Ea Three-Way
$265.00USD / Each
Cadence Luer Stopcock, Nickel-Plated Brass, Three-Way Y-Stopcock, Female Luer x Female Luer x Male Luer Lock EW-41507-15 Mfr # 6056IND
COMPARE
Luer Stopcock Fitting, Nickel-Plated Brass, Three-Way Y-Stopcock, Female Luer x Female Luer x Male Luer Lock; 1/Ea Three-Way
$486.00USD / Each
Showing 1- 8 of 8