This product is now discontinued, View Related Products

Cadmium Nitrate Tetrahydrate

  • Insoluble Matter - ≤0.005%
  • Chloride - ≤0.001%
  • Sulfate - ≤0.003%
  • Ammonia - ≤0.010%
  • Copper - ≤0.002%
  • Iron - ≤0.001%
  • Lead - ≤0.005%
  • Zinc - ≤0.05%
  • Calcium - ≤0.01%
  • Potassium - ≤0.01%
  • Sodium - ≤0.03%
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical C13100 Cadmium Nitrate Tetrahydrate (Cryst./Certified) (100g) EW-88068-79 Mfr # C13100
COMPARE
10022-68-1 Certified
Call For Price
Fisher Chemical C13500 Cadmium Nitrate Tetrahydrate (Cryst./Certified) (500g) EW-88068-82 Mfr # C13500
COMPARE
10022-68-1
Call For Price
Acros Organics AC212342500 Cadmium nitrate tetrahydrate p.a. (250g) EW-88227-08 Mfr # AC212342500
COMPARE
10022-68-1
Call For Price
Acros Organics AC403755000 Cadmium nitrate tetrahydrate, reagent, crystals 99% (500g) EW-88341-34 Mfr # AC403755000
COMPARE
10022-68-1
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat