Cal Control 3300- and 9400-Series Temperature/Process Controllers
Autotuning PID industrial controllers
  • Set points are limited to maximum/minimum temperature ranges for increased safety
  • IP66- and NEMA 4X-rated faceplate allows the controller to be used in washdown applications
  • Retrofittable communication RS232 or RS485
  • Available in 1/32- and 1/16-DIN sizes
  • Input type: Thermocouple (J, K, T, E, N, B, R, S, L), 100 Ω Pt RTD, 5 to 50 mV
These controllers are designed for ease of use with simple menu-driven programming and reliability in demanding applications. They are widely used in many applications such as plastics, packaging, drying, ovens and furnaces, laboratory and scientific equipment. The autotune feature automatically selects the optimal PID values making PID control simple. The unique dAC function is designed to minimize overshoot associated with conventional PID control. The Debug feature checks your communications wiring to produce a visual indication of proper working conditions. These controllers also feature a single ramp/soak (dwell) program, five alarm modes, multilevel security lockout, and heat-cool operation. A mounting sleeve allows for easy mounting, maintenance and field communication retrofits. Each controller shows both PV (process value) and SP (set value) readings.
Call our Applications Specialists for more information on these products or to special order a controller with other options.
$352.00 - $770.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
DIN Size On / Off Function PID Function
Availability Pricing
West Controls 330000000 1/32-DIN Temp. controller with single-line display, SSRD, relay, 100-240 VAC EW-89819-97 Mfr # 330000000
COMPARE
$352.70USD / Each
West Controls 331100000 1/32-DIN Temp. controller with single-line display, relay, relay, 100-240 VAC EW-89819-98 Mfr # 331100000
COMPARE
$495.30USD / Each
West Controls 332200000 1/32-DIN Temp. controller with single-line display, SSRD,SSRD, 100-240 VAC EW-89819-99 Mfr # 332200000
COMPARE
$352.00USD / Each
Cal Control 330000030 1/32-DIN Temp. controller with single-line display, SSRD, relay, 12-24 VDC EW-89820-07 Mfr # 330000030
COMPARE
$386.90USD / Each
West Controls 330 000 230 1/32-DIN controller with single-line display, RS-232, SSRD, relay, 12-24 VDC EW-89820-12 Mfr # 330 000 230
COMPARE
$445.20USD / Each
West Controls 330 000 430 1/32-DIN controller with single-line display, RS-485, SSRD, relay, 12-24 VDC EW-89820-16 Mfr # 330 000 430
COMPARE
$770.00USD / Each
West Controls 940000000 1/16-DIN Temp. controller with dual-line display, SSRD, relay, 100-240 VAC EW-89820-35 Mfr # 940000000
COMPARE
$514.00USD / Each
West Controls 941100000 1/16-DIN Temp. controller with dual-line display, relay, relay, 100-240 VAC EW-89820-37 Mfr # 941100000
COMPARE
$422.40USD / Each
West Controls 940 000 400 1/16-DIN controller with dual-line display, RS-485, SSRD, relay, 100-240 VAC EW-89820-42 Mfr # 940 000 400
COMPARE
$506.25USD / Each
Showing 1- 9 of 9