This product is now discontinued, View Related Products
Caliper electrodes
The BTX Caliper Electrodes are in vivo style re-usable electrodes that are used for a variety of applications. Caliper Electrodes come in two models, with each model consisting of a caliper and a pair of end plates. Model 0289438 are used for smaller animals with a smaller surface area, while model 0289440 is used to target slightly larger surface areas. The end plates of the Caliper Electrode may be adjusted by using the black roller mounted on the caliper. Caliper Electrodes may be cleaned with a mild detergent and sterilized with ethanol or ethylene oxide. Properly maintained Caliper Electrodes have an unlimited life, and are compatible with most BTX Electroporation Systems.
Caliper Electrodes are made of stainless steel and may be used for many electroporation applications, including drug and gene delivery. The electrodes span the target area following injection of the molecule of interest, and electroporation pulses are delivered using a BTX Electroporation System. The electric field introduced by the Caliper Electrodes cause transient pores to form in the cells of the tissue, allowing uptake of the molecules into the cells.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
High Voltage Mode - Capacitance High Voltage Mode - Pulse Length High Voltage Mode - Voltage
Availability Pricing
BTX 45-0101 Electroporation Caliper Electrode, 1 cm x 1 cm EW-02894-38 Mfr # 45-0101
COMPARE
Call For Price
BTX 45-0102 Electroporation Caliper Electrode, 1.5 cm x 1.5 cm EW-02894-40 Mfr # 45-0102