Carbon Block Cartridges for "Big Blue" Housing
The bonded powdered activated carbon filter uses a design to remove organic tastes and odors, chlorine and sediment. Ideal for low or highly variable flow applications since flow cannot channel through the media like granular carbon cartridges††. Cryptosporidium and Giardia cysts are also removed with the 0.5-micron design.
Cartridges are made from FDA compliant materials and meet NSF Standard 42 for materials.
$64.00 - $197.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Length (in) Flow Rate Micron Rating
Availability Pricing
Big Blue Filter cartridge, "", 10", carbon block, 0.5 micron EW-01508-89 Mfr # 155170-43(CBC-BB)
COMPARE
10 2 GPM 0.5
$107.00USD / Each
Big Blue Filter cartridge, "", 20", carbon block, 0.5 micron EW-01508-98 Mfr # 155312-43(CBC-20BB)
COMPARE
20 4 GPM 0.5
$197.00USD / Each
Big Blue Filter cartridge, 10", carbon block, 5 micron EW-01512-54 Mfr # 155548-43(EP-BB)
COMPARE
10 2 GPM 5
$64.00USD / Each
Big Blue Filter cartridge, 20", carbon block, 5 micron EW-01512-56 Mfr # 155583-43(EP-20BB)
COMPARE
20 4 GPM 5
$120.00USD / Each
Showing 1- 4 of 4
Need Help with this product?
Call Us At1-847-549-7600
Live chat