Casella dBadge2® Personal Noise Dosimeters and Kits
The simplest way to obtain and record personal noise exposure
  • Transmit sound level measurements wirelessly to increase productivity
  • Monitor multiple employees' noise exposure remotely and simultaneously
  • Select the appropriate hearing protection equipment—measurements are compliant with various regulation agencies
  • Securely mounts to the shoulder for worry-free, hands-free monitoring
  • Have confidence in your measurements—integral motion sensor expresses motion index in % of time
Transmit sound level measurements through Bluetooth® 4.0 wireless connectivity to increase productivity—no need to stop working to download readings! Dose, status, level alerts and more can be accessed through the free Airwave™ app and sent directly to your Apple or Android enabled device. The dBadge2 noise dosimeters capture every possible noise exposure related parameter in a single measurement run, and make it available for subsequent download and analysis. Email summary data, photographs, and even notes for review or inclusion in reports.
Ensure the correct hearing protection is chosen for each application and location. The measurements taken are in accordance with CFR 1910.95, ISO 9612:2009, and UK Guidance document L108. The LCD displays current noise levels in color-coded format for easy interpretation of alarm conditions at a glance. Use the preset alarm limits to adhere to OSHA, ISO, MSHA, and ACGIH guidelines, or set your own.
Each personal noise dosimeter comes with two mounting clips, one at the top and one at the bottom, for secure shoulder attachment and features a reliable and robust 1/2" diameter microphone with windscreen. Order a kit to get started and add additional units as needed to cover an entire fleet!
$1,860.00 - $21,700.00USD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
Compareitem specifications
Decibel (Db) High Range (dB) Decibel (Db) Low Range (dB) Description
Availability Pricing
Casella DBadge2 Noise Dosimeter; Meter Only EW-50536-51 Mfr # DBADGE2
COMPARE
140.3 54 Noise Dosimeter; Meter Only
$1,860.00USD / Each
Casella DBadge2 Noise Dosimeter Kit with 1 Meter EW-50536-52 Mfr # DBADGE2/K1
COMPARE
140.3 54 Noise Dosimeter Kit with 1 Meter
$2,910.00USD / Each
Casella DBadge2 Noise Dosimeter Kit with 3 Meters EW-50536-53 Mfr # DBADGE2/K3
COMPARE
140.3 54 Noise Dosimeter Kit with 3 Meters
$5,630.00USD / Each
Casella DBadge2 Noise Dosimeter Kit with 5 Meters EW-50536-55 Mfr # DBADGE2/K5
COMPARE
140.3 54 Noise Dosimeter Kit with 5 Meters
$8,060.00USD / Each
Casella DBadge2 Noise Dosimeter Kit with 10 Meters EW-50536-59 Mfr # DBADGE2/K10
COMPARE
140.3 54 Noise Dosimeter Kit with 10 Meters
$13,850.00USD / Each
Casella DBadge2Plus Noise Dosimeter; Meter Only EW-50536-61 Mfr # DBADGE2PLUS
COMPARE
140.3 54 Noise Dosimeter; Meter Only
$2,430.00USD / Each
Casella DBadge2Plus Noise Dosimeter Kit with 1 Meter EW-50536-62 Mfr # DBADGE2PLUS/K1
COMPARE
140.3 54 Noise Dosimeter Kit with 1 Meter
$3,550.00USD / Each
Casella DBadge2Plus Noise Dosimeter Kit with 3 Meters EW-50536-63 Mfr # DBADGE2PLUS/K3
COMPARE
140.3 54 Noise Dosimeter Kit with 3 Meters
$7,250.00USD / Each
Casella DBadge2Plus Noise Dosimeter Kit with 5 Meters EW-50536-65 Mfr # DBADGE2PLUS/K5
COMPARE
140.3 54 Noise Dosimeter Kit with 5 Meters
$10,600.00USD / Each
Casella DBadge2Plus Noise Dosimeter Kit with 10 Meters EW-50536-69 Mfr # DBADGE2PLUS/K10
COMPARE
140.3 54 Noise Dosimeter Kit with 10 Meters
$17,400.00USD / Each
Casella DBadge2Pro Noise Dosimeter; Meter Only EW-50536-71 Mfr # DBADGE2PRO
COMPARE
140.3 54 Noise Dosimeter; Meter Only
$3,080.00USD / Each
Casella DBadge2Pro Noise Dosimeter Kit with 1 Meter EW-50536-72 Mfr # DBADGE2PRO/K1
COMPARE
140.3 54 Noise Dosimeter Kit with 1 Meter
$4,170.00USD / Each
Casella DBadge2Pro Noise Dosimeter Kit with 3 Meters EW-50536-73 Mfr # DBADGE2PRO/K3
COMPARE
140.3 54 Noise Dosimeter Kit with 3 Meters
$8,730.00USD / Each
Casella DBadge2Pro Noise Dosimeter Kit with 5 Meters EW-50536-75 Mfr # DBADGE2PRO/K5
COMPARE
140.3 54 Noise Dosimeter Kit with 5 Meters
$12,900.00USD / Each
Casella DBadge2Pro Noise Dosimeter Kit with 10 Meters EW-50536-79 Mfr # DBADGE2PRO/K10
COMPARE
140.3 54 Noise Dosimeter Kit with 10 Meters
$21,700.00USD / Each
Showing 1- 15 of 15
Need Help with this product?
Call Us At1-847-549-7600
Live chat