Cast iron float and thermostatic steam traps (parallel configuration)
Float and thermostatic steam traps contain a float valve mechanism which modulates to discharge condensate continuously at steam temperature, while non-condensable gases are released by a separate internal balanced pressure thermosatic air vent. Stainless steel internal parts can be serviced without disturbing the piping connections.
Typical applications:
  • All process equipment, particularly when controlled by modulating temperature control valves
  • Unit heaters
  • Air heat coils
  • Heat exchangers
  • Steam main drip stations
$362.50 - $603.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
Spirax Sarco FT-15 3/4" 50926  Cast Iron Float/Thermostatic Steam Trap, Parallel Config.; 3/4"NPT(F), 15 psig EW-07600-30 Mfr # FT-15 3/4" 50926
COMPARE
Cast iron
$362.50USD / Each
Spirax Sarco FT-30 3/4" 50928  Cast Iron Float/Thermostatic Steam Trap, Parallel Config.; 3/4"NPT(F), 30 psig EW-07600-32 Mfr # FT-30 3/4" 50928
COMPARE
Cast iron
$401.10USD / Each
Spirax Sarco FT-75 3/4" 50930  Cast Iron Float/Thermostatic Steam Trap, Parallel Config.; 3/4"NPT(F), 75 psig EW-07600-34 Mfr # FT-75 3/4" 50930
COMPARE
Cast iron
$595.75USD / Each
Spirax Sarco FT-125 3/4" 50932  Cast Iron Float/Thermostatic Steam Trap, Parallel Config.; 3/4"NPT(F), 125 psig EW-07600-36 Mfr # FT-125 3/4" 50932
COMPARE
Cast iron
$603.00USD / Each