Cast Iron Self-Priming Petroleum Centrifugal Pumps
Compatible with three distinct fuel classifications
  • Models for the following fluid groups:
    - Gasoline, kerosene, avgas, and jet fuel
    - Biodiesel, fuel oil, and diesel
    - Ethanol and E85 blend
  • Available in both self-priming and standard end suction configurations
  • Custom configurations are a phone call away to our Application Specialists
These close-coupled pumps have been specifically engineered for compatibility with clean, nonabrasive petroleum products. Transfer and delivery of various fuels such as gasoline, ethanol, biodiesel, and fuel oils are just a few of the petroleum-based products these pumps can handle.
$1,284.00 - $4,435.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Head (ft) Max Motor RPM (rpm)
Availability Pricing
COMPARE
125 54
$2,514.50USD / Each
COMPARE
150 96
$3,331.50USD / Each
COMPARE
150 96
$1,284.00USD / Each
COMPARE
150 96
$1,697.50USD / Each
COMPARE
150 96
$2,695.00USD / Each
COMPARE
215 111
$4,435.00USD / Each