Catalyst FH-Series Multichannel Peristaltic Pump Cartridges
Use with Catalyst multichannel cartridge pump heads for synchronous, multichannel pumping
  • Cartridges feature accurate flow rates and adjustable tubing retainers to hold tubing in place
All cartridges have adjustable occlusion which allows for bi-directional flow variation of ±5%. Tighten occlusion for higher pressures, loosen for longer tubing life. Combine the input/output of two cartridges to reduce pulsation.
$119.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Multichannel Small Cartridge for FH100M 8/3 or 8/4 Pumps
$119.00USD / Each
COMPARE
Multichannel Small Cartridge for FH100M 4/6, 4/8, 12/6 or 12/8 Pumps
$119.00USD / Each